UHP nitrogen generator, NG5000A
Lieferant: PEAK SCIENTIFIC INSTRUMENTS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
10-1295
PEAK10-1295EA 13230 EUR InStock
PEAK10-1295
UHP nitrogen generator, NG5000A
Gasgeneratoren Stickstoff-Generatoren
The NG5000(A) delivers ultra high purity nitrogen at trace detection limits through Pressure Swing Adsorption technology at flows of up to 5 L/min for GC, DSC, DMA and other applications. The NG5000(A) comes with an internal compressor for laboratories without an external source of gas.
  • Suitable for applications such as GC, DSC, DMA
  • Suitable for applications requiring ultra high purity
  • Available with or without an external compressor
  • Contains self-regenerating Carbon Molecular Sieve column
  • Gas is supplied on demand so generator works to your schedule
  • 1 year comprehensive on-site warranty
These generators use Pressure Swing Adsorption and Carbon Molecular Sieve and remove oxygen and other impurities in air, delivering hassle-free, ultra high purity nitrogen, on-demand for the lab.
Outlet pressure: 5,5 bar
Weight: 88 kg
Zertifizierungen: Conforms with: Low Voltage Directive 2014/35/EU; EN 61010-1: 2010 Electrical Equipment for measurement, control and laboratory use.
Electromagnetic Compatibility Directive 2014/30/EU EN 61326-1: 2013 Electrical Equipment for measurement, control and laboratory use.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge