Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Nitrogen generators, precision nitrogen trace

Lieferant: PEAK SCIENTIFIC INSTRUMENTS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

62-0600 62-0250 62-1000
564-3001EA 8680 EUR InStock
564-3001 564-3000 PEAK62-1000
Nitrogen generators, precision nitrogen trace
Gasgeneratoren Stickstoff-Generatoren
These precision nitrogen trace generators produce "zero nitrogen" on demand from a compressed air source.

  • Suitable for carrier gas, make up gas and sample preparation
  • Regenerative CMS columns remove oxygen and moisture
  • Catalyst chamber to remove hydrocarbons (as methane) to <0,05 ppm with no expensive catalyst chamber replacements

Maximum pressure: 5,5 bar

Min/ max operating temperature: 5 to 35 °C

Gas outlet: 1×¹/₄" BSPP

Purity: >99,9995%

Weight: 21 kg (26 kg for the 1,00 l/min flow rate model)

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge