Pressure regulator
Lieferant: PERKIN ELMER GC INSTRUMENTS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
02400085
PERC02400085EA 1988 EUR InStock
PERC02400085
Pressure regulator
Druckminderer
Two-stage regulator for precise outlet pressure control of helium gas. Made of brass body and stainless steel diaphragm for long life and minimum maintenance.
Maximum inlet pressure of 20700 KPa.
Approximate weight: 1,3 kg
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge