Sample cups for analysers
Lieferant: PERKIN ELMER GC INSTRUMENTS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
B0510397 B3001566 B3001567 B0087600 B0119079
PERCB0510397EA 209 EUR InStock
PERCB0510397 PERCB3001566 PERCB3001567 PERCB0087600 PERCB0119079
Sample cups for analysers
Probenbehälter Probengefäße für Analyser
Disposable.
These sample cups are of heavy-duty construction to prevent cracking or tipping. Their conical design allows for use of nearly the entire sample. For concentrated acids including HF for analyses.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge