Peristaltic pump tubing
Lieferant: PERKIN ELMER GC INSTRUMENTS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
B3140730
PERCB3140730EA 139 EUR InStock
PERCB3140730
Peristaltic pump tubing
Schläuche Transferschlauch
Pharmed tubing.
  • High fatigue strength under repeated reversed bending stresses
  • Non-toxic, biocompatible
  • Very low gas permeability
  • Well suited to acids and bases
Used for ICP-OES and ICP-MS applications.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge