Deuterium lamp, TruLife™ 1,5
Lieferant: PERKIN ELMER GC INSTRUMENTS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
L6022728
PERCL6022728EA 1160 EUR InStock
PERCL6022728
Deuterium lamp, TruLife™ 1,5
Beleuchtung Deuterium-Lampen
These deuterium lamps provide the best wavelength coverage across the working range of the spectrometer for consistently high performance.
  • Lamps are pre-aligned for easy installation
  • Extended lifetime of 1500 hours
  • Built-in digital timer to monitor lamp lifetime
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge