Multi-element standards, 10 components in 2% HNO₃
Lieferant: PERKIN ELMER GC INSTRUMENTS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
N0691579
PERCN0691579EA 267 EUR InStock
PERCN0691579
Multi-element standards, 10 components in 2% HNO₃
Standards Kationischer Standards Mehrelement-Standards
1 mg/l of barium, magnesium
10 mg/l of lanthanum, lithium, manganese, nickel, strontium, zinc
50 mg/l of arsenic, potassium
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge