Vial racks, for Cetac autosampler
Lieferant: PERKIN ELMER GC INSTRUMENTS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
N0774085 N0777151 N0777153 N0777154 N0777298
PERCN0774085EA 532 EUR InStock
PERCN0774085 PERCN0777151 PERCN0777153 PERCN0777154 PERCN0777298
Vial racks, for Cetac autosampler
Gestelle Gestelle für Chromatographie-Ampullen
These polypropylene racks accommodate 40 headspace auto-sampler vials.
  • Autoclavable and stackable
  • Good chemical and temperature resistance
  • Features end plates for labeling and numbering for tube identification
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge