Multi-element calibration standard 5
Lieferant: PERKIN ELMER GC INSTRUMENTS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
N9300235
PERCN9300235EA 221 EUR InStock
PERCN9300235
Multi-element calibration standard 5
Standards Kationischer Standards Mehrelement-Standards
Designed to contain virtually every element in the mass spectrum for concentration checks.
Concentration of element includes 10 µg/ml of boron, germanium, molybdenum, niobium, phosphorus, rhenium, sulphur, silicon, tantalum, titanium, tungsten, and zirconium.
These solutions are made with the highest purity materials available and are tested on the ELAN 6000 under an internationally accredited ISO 9001 quality assurance program.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge