Inline gas filters
Lieferant: PERKIN ELMER GC INSTRUMENTS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
N9301178
PERCN9301178EA 440 EUR InStock
PERCN9301178
Inline gas filters
Filter Gasfilter
This in-line filter removes moisture, oil, and particulate from gas supplies. It consists of a molecular sieve and indicator in a clear acrylic tube. The drierite changes colour at low relative humidity indicating that packing must be changed.
  • Large capacity filter requires fewer filter changes
  • Improves baseline stability and chromatographic results
Max. pressure: 6,89 bar
Dimensions (W×H): 57×432 mm with 1/8” fittings
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge