Mercury standard solution, for ICP-MS, Pure Plus grade
Lieferant: PERKIN ELMER GC INSTRUMENTS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
N9303740
PERCN9303740EA 93.1 EUR InStock
PERCN9303740
Mercury standard solution, for ICP-MS, Pure Plus grade
Standards Kationischer Standards Quecksilber-Standards
Single element standard is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.
  • Analysed by ICP-OES
  • Analysed by classical wet assay
  • 67 trace impurities analysed by ICP-MS of the final solution and reported on the certificate
  • Impurities reported at ppb level
  • Prepared and certified under ISO Guide 34 and ISO 17025 – certified by A2LA
Achtung! Bitte beachten: These standards are in acid solution, for full details of hazards please read the accompanying Material Safety Data Sheets (MSDS).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge