Calibration standard, single element, Lithium for AAS and ICP-OES, pure grade
Lieferant: PERKIN ELMER GC INSTRUMENTS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
N9303781
PERCN9303781EA 69.6 EUR InStock
PERCN9303781
Calibration standard, single element, Lithium for AAS and ICP-OES, pure grade
Standards Kationischer Standards Lithium-Standards
Pure Grade Standards for AA and ICP-OES
  • Analyzed by ICP-OES
  • Analyzed by Classical Wet Assay
  • 32 trace impurities analyzed by ICP-MS of the final solution and reported on the certificate
  • Impurities reported at ppm level
  • All Standards are prepared and certified under ISO Guide 34 and ISO 17025 – certified by A2LA
Achtung! Bitte beachten: These standards are in acid solution, for full details of hazards and specification, please see website and the links to Material Safety Data Sheets (MSDS) and Technical Data sheets.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge