Push brooms, stiff sweep
Lieferant: PERFEX
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2524B
PERF2524BEA 207 EUR InStock
PERF2524B
Push brooms, stiff sweep
Besen
These brooms are made of extremely durable polypropylene fibres which are non-conductive and will not absorb bacteria, liquids, or odours.
This durable fill is excellent for sweeping heavy or coarse material and for removing difficult dirt and deposits from unfinished surfaces, wood, concrete, blacktop, etc.
Bestellinformation: Polymer adjustable or fixed length handles must be ordered separately.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge