Recirculating chiller for ICP/ICP-MS
Lieferant: PerkinElmer
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
N0772025
PERKN0772025EA 0 EUR InStock
PERKN0772025
Recirculating chiller for ICP/ICP-MS
Umwälzthermostate Geschlossene Umwälzsysteme Kälteumwälzthermostate
Water cooling systems provide clean, reliable temperature-controlled fluid for open tanks or closed loop cooling and are ideal for many routine process cooling needs.
  • Feature convenient 'one-touch' temperature control and bright LED readouts
  • Instant alerts through audible and visual alarms when operating parameters exceed pre-set limits
  • Built-in safeties to alert low fluid levels and thermal runaway
Other features include a calibration function that allows to adjust the temperature display to match a traceable temperature standard and optional interfaces for various control functions, such as on/off and remote control.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge