Wipes for delicate tasks, VERACLEAN® I-Tack
Lieferant: PGI NONWOVEN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
74236
PGIN74236EA 325 EUR InStock
PGIN74236
Wipes for delicate tasks, VERACLEAN® I-Tack
Wischtücher Wisch- und Handtücher, Einweg Wischtücher für sensible Anwendungen
These wipes are specially designed to swiftly and cleanly remove contaminants prior to painting. This I-Tack range contains a water-based tackifier and is compatible with all paint types.
  • The cloths are cost efficient because they can be used up to eight sides
  • No unpleasant smells or sticky residue on either your hands or the car body
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge