Microtubes, 1,1 ml
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
15088
PIER15088EA 59.5 EUR InStock
PIER15088
Microtubes, 1,1 ml
Röhrchen Mikroröhrchen
Microtube racke system consists of 1,1 ml-tall microtubes supplied pre-racked in boxes to match a standard 96-well microplate footprint.
  • Available individually or in an 8×12 microtitration plate format
  • 8×12 format simplifies and speeds applications in which a multi-channel pipettor is used
  • Format is compatible with robotic workstations
  • Microtube system can be centrifuged
The microtubes resemble 1 ml pipette tips, except that they have round bottoms. Like a box of 1 ml pipette tips, the racked microtubes match multi-channel pipettors, providing convenience for a variety of reagent- and sample-dispensing procedures.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge