West HisProbe kit, SuperSignal™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
15168
PIER15168EA 696 EUR InStock
PIER15168
West HisProbe kit, SuperSignal™
Elektrophorese-Reagenzien Western Blotting Reagents
SuperSignal™ West HisProbe Kit contains HisProbe-HRP conjugate and reagents required for detection of polyhistidine-tagged protein by Western blotting. HisProbe-HRP Conjugate is a nickel (Ni²⁺)-activated derivative of horseradish peroxidase (HRP) that enables direct, IMAC-based detection of His-tagged proteins.
  • One-step probe incubation eliminates the need to run a lengthy two-step primary/secondary antibody reaction protocol
  • Recognise polyhistidine tags independent of adjacent tags
  • Smaller than antibodies and detects his tagged fusion proteins that are often undetectable
The active ligand in HisProbe-HRP is a tridentate chelator that allows Ni²⁺ to be bound in active form for subsequent interaction and detection of target molecules. The active chelator has binding capabilities similar to iminodiacetic acid (IDA) when used for immobilised metal affinity chromatography (IMAC).
Lieferumfang: Kit includes HisProbe-HRP conjugate (2 mg), SuperSignal™ West Pico Luminol/Enhancer solution (250 ml), SuperSignal™ West Pico stable peroxide solution (250 ml), Blocker BSA in TBS, 10X (125 ml), TBS packs (10 packs, each making 500 ml), Surfact-Amps™ 20 detergent solution (6×10 ml).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge