Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Boronic acid resin

Lieferant: Thermo Scientific

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


20244
PIER20244EA 327 EUR InStock
PIER20244
Boronic acid resin
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
Pierce™ Boronic acid resin enables ribonucleotide and oligonucleotide RNA isolation.

  • Support: Beaded polyacrylamide (1800 MW exclusion limit)
  • Spacer: m-aminophenyl group
  • Loading: 100 µMol boronate per ml of settled resin
  • Capacity: at least 99% binding and recovery of 110 µMol AMP per ml of settled resin
  • Format: 50% resin slurry in 0,05% sodium azide

The immobilised boronic acid is an easy to use polyacrylamide affinity support for purification of ribonucleotides and other small molecular weight compounds that contain cis-diol groups. Boronic acid affinity chromatography has been used to isolate ribonucleoside, to purify nucleosidyl peptide, separate ribonucleosidases in tissue extracts, separate RNA and oligoribonucleotide, and determine of the amount of non-enzymatic glycosylation present in peripheral nerve from diabetic and control rats and dogs.


Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge