Coupling resin, CarboxyLink™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
20266
PIER20266EA 267 EUR InStock
PIER20266
Coupling resin, CarboxyLink™
Protein-Aufbereitungs Protein Purification Resins
CarboxyLink™ coupling resin is used for covalent immobilisation of peptides or other carboxyl-containing molecules to a porous, beaded resin for use in affinity purification procedures.
  • DADPA-activated crosslinked 4% beaded agarose
  • Couple 1 to 2 mg of peptide per ml of resin
  • Immobilises peptides with high capacity and various orientations without steric hindrance
  • Immoblisation results in covalent attachment of carboxyl groups by amide bonds, allowing for multiple rounds of affinity purification with one batch of prepared resin
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge