Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Affinity purification beads and magnets, MagnaBind™

Lieferant: Thermo Scientific

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

21354 21356 21344 21352 21353 21349 21348
PIER21354EA 506 EUR InStock
PIER21354 PIER21356 PIER21344 PIER21352 PIER21353 PIER21349 PIER21348
Affinity purification beads and magnets, MagnaBind™
Kugeln Magnetkugeln
Magnetic beads are a convenient affinity support for a variety of assays, which allow easy purification of the target without columns or centrifugation. After a binding step in an affinity purification procedure, the magnetic particles are easily and rapidly collected by placing the microcentrifuge tube or reaction vessel next an appropriate rare-earth magnet. MagnaBind™ beads respond rapidly to MagnaBind™ magnets but can be easily dispersed and regathered multiple times (ie. they will not irreversibly aggregate) because they do not have any magnetic memory.

  • Available pre-coated with popular affinity ligands or derivatised for covalent attachment of proteins and other specific ligands
  • Beads do not irreversibly aggregate because they have no magnetic memory; collect and disperse the beads multiple times, if needed
  • Most separations require a short 5 to 10 minute bench-top procedure

MagnaBind™ beads are available pre-coated with Protein A, Protein G, streptavidin, anti-mouse or anti-rabbit antibodies. Activated beads, with either amine or carboxyl groups, are also available for attaching other proteins or affinity ligands to a magnetic particle.

These beads are applicable in cell sorting using either positive or negative selection and protein purification or immunoassays using either direct or indirect methods.

Features of MagnaBind beads:
Composition: Silanized iron oxide
Magnetisation: 25 to 35 EMU/g
Type of magnetisation: Superparamagnetic (no magnetic memory)
Surface area: >100 m²/g
Bead size: 1 to 4 µm diameter
Settling rate: 4% in 30 minutes
Effective density: 2,5 g/ml
Number of beads: 1×10⁸ beads/mg
pH stability: Aqueous solution, above pH 4,0

Achtung! Bitte beachten: For research use only. Not for use in diagnostic procedures.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge