Pierce™ 660 nm Protein Assay Kit and Reagent
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
22660 22662
PIER22660EA 228 EUR InStock
PIER22660 PIER22662
Pierce™ 660 nm Protein Assay Kit and Reagent
Tests Proteintests
Pierce™ 660 nm Protein Assay is a ready to use, detergent- and reducing agent-compatible assay reagent to quickly measure (A660 nm) total protein concentration compared to a protein standard.
  • Can be performed in either a test tube or microplate format
  • Works with wide range of detergents and reducing agents than other dye-based assays
  • Single reagent with a simple mix-and-read protocol; no working reagent to prepare
  • Requires only 10 µl for microplate or 100 µl for the test tube procedures
Pierce™ 660 nm Assay is more linear than Coomassie-based Bradford assays and compatible with higher concentrations of most detergents, reducing agents and other commonly used reagents. The accessory Ionic Detergent Compatibility Reagent (IDCR) provides for even broader detergent compatibility, making this one of the only protein assays that is suitable for samples containing Laemmli SDS sample buffer with Bromophenol blue.
Lieferumfang: Kit includes Pierce™ 660 nm Protein Assay Reagent (450 ml) and albumin standard pre-diluted set (7×3,5 ml). Pierce™ 660 nm Protein Assay Reagent is also available separately.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge