Compat-Able™ Protein assay preparation reagent set
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
23215
PIER23215EA 176 EUR InStock
PIER23215
Compat-Able™ Protein assay preparation reagent set
Tests Proteintests
The Compat-Able™ Protein assay preparation reagent set eliminates interfering substances from protein assay samples to make them compatible with BCA and Bradford assays.
  • Removes nearly any substance that interferes with colorimetric protein assays: salts, detergents, denaturants, reducing agents, and even IEF sample buffer
  • Prepare samples in less than 10 minutes with four simple steps
  • Reagent set is easier to use than homemade TCA or acetone precipitation reagents, thereby producing more consistent results
  • Microcentrifuge method is easily scaled to larger centrifuge tube volumes for special needs
  • Protocol integrates control and normalisation procedures
Lieferumfang: Kit contains sufficient reagents to pretreat up to 500 samples. It comprises Compat-Able™ Protein assay preparation reagent 1 (250 ml) and Compat-Able™ Protein assay preparation reagent 2 (250 ml).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge