Pierce™ Coomassie Plus (Bradford) Protein Assay Kit
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
23236
PIER23236EA 308 EUR InStock
PIER23236
Pierce™ Coomassie Plus (Bradford) Protein Assay Kit
Tests Proteintests
The Pierce™ Coomassie Plus Protein assay is a quick and ready-to-use, reducing agent-compatible, improved Bradford assay to quickly measure (A595 nm) total protein concentration compared to a protein standard.
  • Colorimetric measure with a standard spectrophotometer or plate reader (595 nm)
  • Easy to use single reagent; no working reagent preparation required
  • Fast almost immediate colour development; add, mix and read results
  • Broad range detects protein concentration in the range 1 to 1500 µg/ml
  • Better improved linearity and response-uniformity compared to traditional Bradford formulations
  • Flexible microplate and cuvette protocols provided with the instructions and adaptable to several target working ranges
This modification of the well-known Bradford method greatly reduces the tendency of Coomassie reagents to give non linear response curves by a formulation that substantially improves linearity for a defined range of protein concentration. In addition, the Coomassie Plus Reagent results in significantly less protein-to-protein variation than is observed with other Bradford-type Coomassie formulations.
Lieferumfang: Kit contains sufficient reagents for 630 tube assays or 3160 microplate assays. Pierce™ Coomassie Plus™ Protein Assay Reagent, 950 ml; Albumin Standard Ampoules, 2 mg/ml, 10×1 ml.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge