Pierce™ Coomassie Plus (Bradford) Protein Assay Reagent
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
23238
PIER23238EA 237 EUR InStock
PIER23238
Pierce™ Coomassie Plus (Bradford) Protein Assay Reagent
Tests Proteintests
The bradford protein assay is the preferred colorimetric assay for quantifying total protein concentration. The Pierce™ coomassie plus protein assay is a ready-to-use, reducing agent-compatible, improved bradford assay reagent to quickly measure (A595 nm) total protein concentration compared to a protein standard.
  • Sensitive, fast and consistent protein assay kit
  • Determine protein concentrations from 1 to 1500 μg/ml
  • Ready-to-use dye binding reagent formulation
  • Convenient microplate or cuvette format
  • Compatible with reducing sugars, reducing substances and thiols
This modification of the well-known bradford method greatly reduces the tendency of coomassie reagents to give non linear response curves by a formulation that substantially improves linearity for a defined range of protein concentration. In addition, coomassie plus reagent results in significantly less protein-to-protein variation than is observed with other bradford-type coomassie formulations.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge