Easy-Titer® IgG assay kits
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
23305 23300 23310 23325
PIEA23305EA 0 EUR InStock
PIEA23305 PIER23300 PIER23310 PIEA23325
Easy-Titer® IgG assay kits
Tests Immunologische Tests
Easy-Titer® IgG assay kits are simple, mix-and-read assays that allow quick and accurate determination of concentrations from 15 to 300 ng/ml of IgG.
  • Faster and easier than ELISA
  • Sensitive, requires small sample volume
  • No special equipment needed
  • Highly reproducible results
The assay procedure uses monodispersed polystyrene beads that are coated with anti-IgG antibodies and absorb light at 340 and 405 nm. When the beads are mixed with a sample containing IgG, they aggregate, causing decreased absorption of light and, therefore, low IgG concentrations yield high absorbance values and high IgG concentrations yield low absorbance values. The decrease in absorption is proportional to IgG concentration and a standard curve can be generated to accurately quantify levels of IgG in serum, ascites or cell culture supernatant samples.
Lieferumfang: Kits include sufficient components for 96 tests. An IgG standard must be purchased separately. Each kit contains either Anti-Mouse, Anti-Rabbit, Anti-Human IgG (H+L) or Anti-Human IgG (gamma chain) sensitised polystyrene beads (2 ml), dilution buffer (30 ml), and blocking reagent (15 ml).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge