Amino acid modification reagents, sulfhydryl addition kit, Pierce™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
23460
PIER23460EA 379 EUR InStock
PIER23460
Amino acid modification reagents, sulfhydryl addition kit, Pierce™
Protein-Aufbereitungs Protein Purification Kits
The Pierce™ Sulfhydryl Addition Kit contains all necessary components to introduce free sulfhydryl groups into any primary amine-containing molecule, purify the modified molecule and quantify the added sulfhydryl groups.
The Sulfhydryl Addition Kit uses SATA (N-succinimidyl S-acetylthioacetate) as a means of adding sulfhydryls to molecules. SATA contains an N-hydroxysuccinimide (NHS) ester that forms a stable amide bond with primary amines. Once the covalent linkage is formed, the modified molecule can be stored and the sulfhydryl group de-protected when needed. The reaction conditions for SATA are mild and non-denaturing, which preserves protein activity.
Lieferumfang: The kit includes SATA (2 mg); hydroxylamine-HCl (5 mg); 10X conjugation buffer stock (20 ml); phosphate buffered saline pack (1 pack); dimethylformamide (DMF, 1 ml); dextran desalting column (1×5 ml); column extender (1); Ellman’s reagent (DTNB, 2 mg); and cysteine-HCl H₂O (20 mg).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge
Diese Webseite verwendet Cookies, eigene oder von Drittanbieter, zur Sammlung statistischer Informationen, um Inhalte und Anzeigen für Sie zu personalisieren. Wenn Sie diese Seite weiterhin verwenden, akzeptieren Sie die Cookie-Nutzung.
Weitere Informationen finden Sie in unseren Datenschutzbestimmungen..