Immunoprecipitation, magnetic ChIP kit, Pierce™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
26157
PIER26157EA 818 EUR InStock
PIER26157
Immunoprecipitation, magnetic ChIP kit, Pierce™
Protein-Aufbereitungs Protein Purification Kits
The Pierce™ Magnetic ChIP Kit provides a convenient method for efficient isolation of chromatin-bound DNA by immunoprecipitation for subsequent quantitation by PCR.
  • Purified DNA for quantitative PCR can be obtained in about 8 hours
  • Optimised micrococcal nuclease digestion and nuclear lysis
  • Obtain results with as few as 10000 cells
The blocked protein A/G magnetic beads provide high binding capacity, low non specific background, and work with many antibody species. These beads can be used manually with a magnetic stand or with automated platforms.
Lieferumfang: The kit contains sufficient reagents to perform 30 chromatin immunoprecipitation (ChIP) assays with appropriate controls using an optimised protocol. The kit includes reagents for cell lysis, capture of protein-DNA complexes, reversal of crosslinking, and DNA isolation.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge