Multicolour protein ladder, broad range, Spectra™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
26634
PIER26634EA 218 EUR InStock
PIER26634
Multicolour protein ladder, broad range, Spectra™
Elektrophorese-Reagenzien Molekulargewichtsmarker
The Spectra™ Multicolour Broad Range Protein Ladder is a 4-colour protein standard containing 10 prestained proteins (10 to 260 kDa) for use in gel electrophoresis and Western blotting.
  • Four different colours (blue, orange, green, pink) for unambiguous band-size assignment
  • Supplied in a loading buffer for direct loading on gels
  • Colour-coded bands of similar intensity for easy visualisation
  • Evaluated by SDS-PAGE and Western blotting
  • Coloured bands transfer to membranes for Western blotting
Spectra™ Multicolour Broad Range Protein Ladder is designed for monitoring the progress of SDS-polyacrylamide gel electrophoresis, for assessing transfer efficiency onto PVDF, nylon and nitrocellulose membranes, and for estimating the approximate size of separated proteins that have been made visible with gel stains or Western blot detection reagents.
Lieferumfang: Kit includes dye-stained proteins in 62,5 mM Tris-H3PO4 (pH 7,5 at 25 °C), 1 mM EDTA, 2% SDS, 10 mM DTT, 1 mM NaN3 and 33% glycerol.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge