28320
28328
28336
28316
28324
28332
28321
28313
28329
28230
85115
85114
85113
85112
85111
85117
85118
85125
PIER28320EA
219
EUR
InStock
PIER28320
PIER28328
PIER28336
PIER28316
PIER28324
PIER28332
PIER28321
PIER28313
PIER28329
PIER28230
PIER85115
PIER85114
PIER85113
PIER85112
PIER85111
PIER85117
PIER85118
PIER85125
Detergent solutions, Surfact-Amps™
Protein-Extraktion
Proteinextraktions-Reagenzien
Surfact-Amps™ detergents are ready-to-use, diluted solutions of purified detergent that can be used in routine and high-demand protein research methods and molecular biology techniques. These detergents are useful as a component of cell lysis buffers or other solutions intended to extract and solubilise proteins, improve chromatography, prepare yeast spheroplasts, and reduce background in ELISA or Western blot methods.
- Clear and pure 10% detergent solutions with <1.0 μeq/ml peroxides and carbonyl content
- Ampoules are packed under nitrogen for maximum shelf life and are pre-scored for easy opening
- Easy to pipette and accurately dispense
- Packaging eliminates the accumulation of peroxides and degradation products
Detergents are a class of molecules whose unique properties enable manipulation (disruption or formation) of hydrophobichydrophilic interactions among molecules in biological samples. In biological research, detergents are used to lyse cells (release soluble proteins), solubilise membrane proteins and lipids, control protein crystallisation and prevent non specific binding in affinity purification and immunoassay procedures. They are often used as an additive in protein gel electrophoresis.