TAE buffer (20X), Pierce™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
28354
PIER28354EA 116 EUR InStock
PIER28354
TAE buffer (20X), Pierce™
Puffer Elektrophorese-Puffer Laufpuffer
20X TAE buffer is a space-saving stock solution that is ideal for casting and preparing Tris-Acetate EDTA running buffer used for agarose gel electrophoresis of nucleic acids.
When diluted to 1X with water, yields 00,4 M Tris, 0,04 M acetate, 1 mM EDTA, pH 8,2 to 8,4.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge