Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Pierce™ Vacuum Hydrolysis Tubes

Lieferant: Thermo Scientific

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

29570 29571 29572
PIER29570EA 252 EUR InStock
PIER29570 PIER29571 PIER29572
Pierce™ Vacuum Hydrolysis Tubes
Röhrchen Reagenzgläser- und Probenröhrchen
These are high quality glass tubes designed for heated, acid-based, vacuum hydrolysis of proteins and peptides for amino acid analysis. It fits conveniently into reacti-block aluminium heating blocks.

  • Fits into Reacti-Block aluminium heating blocks
  • Sufficient for samples up to 0,35 ml
  • Optimal hydrolysis control

The upper temperature limit of the vacuum hydrolysis tubes is 260 °C, however, do not heat the tubes greater than 100 °C in an oven.

Order Now
 

PRODUKT

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge