Substrate for western blotting, NBT (nitro-blue tetrazolium chloride)
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
34035
PIER34035EA 218 EUR InStock
PIER34035
Substrate for western blotting, NBT (nitro-blue tetrazolium chloride)
Elektrophorese-Reagenzien Western Blotting Reagents
NBT (nitro-blue tetrazolium chloride) yields a blue-purple, water-insoluble product upon reduction of the tetrazolium salt. The product of reduction is NBT-formazan. This substrate is widely used for immunochemistry applications because the color is linear and stable. NBT is used as the substrate for both glucose oxidase and alkaline phosphatase systems.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge