Pierce™ Peroxidase Suppressor
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
35000
PIER35000EA 191 EUR InStock
PIER35000
Pierce™ Peroxidase Suppressor
Tests Immunologische Tests
This peroxidase suppressor is a stable and easy-to-use inhibitor of endogenous peroxidase activity from cells and tissues.
  • Stable and ready-to-use
  • Effectively inhibits peroxidase activity
  • No interference with antigen-antibody interactions
Designed to reduce background signal in HRP-based immunohistochemistry applications.
Achtung! Bitte beachten: Store at 4 °C upon receipt.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge