Peroxidase IHC detection kit, Pierce™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
36000
PIER36000EA 568 EUR InStock
PIER36000
Peroxidase IHC detection kit, Pierce™
Tests Immunologische Tests
The Pierce™ Peroxidase IHC Detection Kit bundles the Pierce™ Metal Enhanced DAB Substrate Kit with all necessary components to stain, counterstain and preserve experimental results with frozen or paraffin tissue sections.
  • Fifty times more sensitive than the traditional DAB (3,3'-diaminobenzidine) method and 30 times more sensitive than other metal-intensified versions of DAB
  • Low background, high intensity, large dynamic range - gives a crisp dark brown-black precipitate that is more intense than the dull brown precipitate observed when using DAB without enhancement; substrate delivers increased intensity with almost non existent background
  • Easy-to-use, two component system
  • Working solution stable for six hours
  • Complete kit
  • Compatible with direct or avidin-biotin-complex (ABC) probing strategies
Lieferumfang: DAB/metal concentrate (1X), 25 ml; Universal Blocker™ blocking buffer in TBS, 25 ml; Peroxidase Suppressor, 2×1 ml; stable peroxide buffer, 25 ml; BupH™ tris buffered saline, 4×5 ml; Surfact-Amps® 2 (1% Tween®-2 Detergent), 1 ml; Harris modified hematoxylin (without acetic acid, mercury-free), 1 ml; and mounting medium, 6 ml.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge