Blocking buffer, Blocker™ BSA (10X) in PBS or TBS
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
37525
PIER37525EA 230 EUR InStock
PIER37525
Blocking buffer, Blocker™ BSA (10X) in PBS or TBS
Puffer Elektrophorese-Puffer Blockier-Puffer
Blocker BSA in PBS or TBS are ready-to-use, 1X PBS or TBS solutions of 10X (10%) bovine serum albumin protein for blocking steps in Western blot, ELISA, IHC and nucleic acid detection methods.
  • Easy to use
  • Convenient and saves storage space
  • Purified protein provides fewer chances of cross reaction with assay components
This 10% (w/v) solution of high quality BSA that is useful for saturating excess protein binding sites on membranes and microplates in immunoassays. Blocker BSA Buffers are most frequently diluted 10-fold (to 1% BSA) in 1X Tris-buffered saline for initial testing, but other buffer concentrations can be beneficial for specific systems. Blocker BSA is usually more effective than non fat milk blocking buffers for biotin-avidin systems because it contains a single purified protein that is devoid endogenous biotin.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge