Blocker™ Blocking Buffer, BLOTTO in TBS
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
37530
PIER37530EA 178 EUR InStock
PIER37530
Blocker™ Blocking Buffer, BLOTTO in TBS
Puffer Elektrophorese-Puffer Blockier-Puffer
Blocker™ BLOTTO is a ready-to-use 5% solution of non fat powdered milk in Tris-buffered saline for Western blot, ELISA, IHC and other detection methods.
  • Convenient as can be diluted as needed
  • Easy to use, formulated with anti-foaming agent and thimerosal-free preservative
  • Flexible may be used for multiple applications
Achtung! Bitte beachten: For research use only. Not for use in diagnostic procedures.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge