Blocking buffer, dry blend, SuperBlock™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
37545
PIER37545EA 296 EUR InStock
PIER37545
Blocking buffer, dry blend, SuperBlock™
Puffer Elektrophorese-Puffer Blockier-Puffer
SuperBlock (TBS) Blocking Buffer is an optimised TBS solution containing a protein that provides incredibly fast and effective blocking for ELISA, IHC and Western blot analysis.
  • Block membranes in 5 to 10 minutes and ELISA plates in 2 minutes
  • Biotin-free for use with streptavidi
  • Available in multiple package sizes
  • Non serum protein solution yields a high signal-to-noise ratio
  • Stable at 4 °C for one year
The protein based formulation does not contain any immunoglobulins, albumin or endogenous biotin, making it compatible in many situations where traditional blocking agents fail. The buffer is especially effective at blocking coated polystyrene microplates (96-well plates) and stabilizing them for drying and storage for later use. Each pack contains a proprietary protein formulation in Tris buffered saline, pH 7.4; each pack/pouch yields 200 ml.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge