North2South™ Hybridisation Buffer
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
37549
PIER37549EA 217 EUR InStock
PIER37549
North2South™ Hybridisation Buffer
Puffer Elektrophorese-Puffer Hybridisierungspuffer
North2South™ hybridisation buffer is an aqueous buffer which is used with the North2South™ chemiluminescent detection kit.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge