Pierce™, CarboxyLink™ Immobilisation Kit
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
44899
PIER44899EA 708 EUR InStock
PIER44899
Pierce™, CarboxyLink™ Immobilisation Kit
Protein-Aufbereitungs Protein Purification Kits
CarboxyLink™ immobilisation kit is for covalent immobilisation of peptides or other carboxyl-containing (—COOH) molecules to a porous, beaded resin for use in affinity purification procedures.
  • Ideal for unmodified peptides, immobilises peptides with high capacity and various orientations without steric hindrance
  • DADPA-activated crosslinked 4% beaded agarose acting as a CarboxyLink™ coupling resin
  • With stable linkage, flexible and gentle coupling conditions
Lieferumfang: Kit includes CarboxyLink™ columns, 2 ml (5 columns), EDC crosslinker (5×60 mg), MES buffered saline pack (makes 500 ml), wash solution (120 ml), and column accessories.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge