Pierce™ Spin Cups - Cellulose Acetate Filter
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
69702
PIER69702EA 202 EUR InStock
PIER69702
Pierce™ Spin Cups - Cellulose Acetate Filter
Filter Zentrifugaleinheiten
Pierce™ cellulose acetate filter spin cups are convenient tools for manipulating small volumes of affinity supports (20 - 400 µl) for protein purification. Simply add the affinity resin and sample to one of the spin cups, then use a microcentrifuge to efficiently wash away contaminants and elute the purified sample without losing any resin in the process. Spin cups allow more protein to be affinity purified in less time.
  • Cellulose acetate filters have smaller pore sizes than paper
  • Column volume: 800 µl
  • Resin volume: 20 to 400 µl
  • Filter type: Cellulose acetate filter, 0,45 µm pore size
  • Cap: Collection tube cap fits onto inserted spin cup
Applications include affinity purification/affinity chromatography, immunodepletion, immunoprecipitation (IP), and co-immunoprecipitation (co-IP).
Format: Microcentrifuge, spin column
Column type: Empty column
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge