Freund's incomplete adjuvant, Imject™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
77145
PIER77145EA 133 EUR InStock
PIER77145
Freund's incomplete adjuvant, Imject™
Protein- Expression Proteinexpressions-Reagenzien
Imject™ Freund's Incomplete Adjuvant (FIA) is a popular non specific stimulator of the immune response for use in antibody production immunisation protocols.
  • Enhances immune response in booster injections
  • Typically used for subsequent boosts after initial injection with FCA
  • Water-in-oil emulsion without Mycobacterium
Lieferumfang: Supplied ready to use in 10 ml ampoules.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge