Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Wasser mit 0,1% (v/v) Ameisensäure ≥99% für die LC-MS, Pierce™

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


85170
PIER85170EA 103 EUR InStock
PIER85170
Wasser mit 0,1% (v/v) Ameisensäure ≥99% für die LC-MS, Pierce™
Wasser mit 0,1% (v/v) Ameisensäure
Pierce™ 0.1% Formic Acid (v/v) in Water is an LC/MS-grade preparation with high purity and low UV-absorptivity that is ideal for HPLC and mass spectrometry applications.

  • Tested more than 20 ways to ensure maximal LC/MS sensitivity
  • Processed, filtered and sealed to extend LC/MS column life
  • Packaged to eliminate variability and reduce handling

This 0.1% Formic Acid in Water is specially purified by a proprietary method and tested to ensure lot-to-lot consistency with a low UV absorbance, providing the most sensitive detection across wavelengths and prolonging equipment life. The chromatography- and spectrometry-grade 0.1% FA in water is 0.2-micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen with PTFE-lined fluorocarbon caps for ultimate protection.

This preparation of Thermo Scientific 0.1% Formic Acid (v/v) in Water is specially purified and tested to the highest specifications to ensure the integrity of chromatography data, maximize assay sensitivity and prolong the life of equipment.

Formic acid is a common component of reverse-phase mobile phases that provide protons for LC/MS analysis. The presence of a low concentration of formic acid in the mobile phase is also known to improve the peak shapes of the resulting separation. Unlike trifluoroacetic acid (TFA), formic acid is not an ion-pairing agent and it does not suppress MS ionization of polypeptides when used as a mobile-phase component.

Achtung! Bitte beachten: For Research Use Only. Not for use in diagnostic procedures.


Storage Temperature: Raumtemperatur
EINECS: 200-579-1
REACH: 01-2119491174-37-XXXX
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=e79a8c7fb6c358f57ed533653859d68b28119045&prid=web&clid=SAPDAM
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge