Graphite spin columns, Pierce™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
88302
PIER88302EA 206 EUR InStock
PIER88302
Graphite spin columns, Pierce™
Filter Zentrifugaleinheiten
Pierce™ Graphite spin columns improve phosphopeptide analysis by efficiently binding hydrophilic peptides and efficiently removing urea, salts and other contaminants before mass spectrometry analysis.
  • Convenient spin format for parallel processing of multiple samples
  • High-binding capacity with recovery of up to 100 µg of hydrophilic peptides per column
  • Porous graphite resin for cleaning up phosphopeptide samples before MS analysis
Enable fast and efficient capture, concentration, desalting and elution of hydrophilic peptides in less than ten minutes.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge