Pierce™ Streptavidin Magnetic Beads
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
88817 88816
PIER88817EA 1310 EUR InStock
PIER88817 PIER88816
Pierce™ Streptavidin Magnetic Beads
Kugeln Magnetkugeln
These Pierce™ streptavidin magnetic beads accelerate throughput for automated magnetic purification of biotinylated molecules.
  • Streptavidin is immobilised using leach-resistant chemistry
  • Superior quality beads with high binding capacity provide rapid and efficient biomolecule purification from complex samples
  • Stable, pre-blocked beads provide clean purification products (eg. antigen eluted in IP with biotinylated antibody) that are compatible with mass spectrometry analysis and other downstream analyses
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge