NHS-Activated magnetic beads, Pierce™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
88826 88827
PIER88826EA 344 EUR InStock
PIER88826 PIER88827
NHS-Activated magnetic beads, Pierce™
Kugeln Magnetkugeln
The Pierce™ NHS-activated magnetic beads enable covalent, amine-based conjugation of proteins to magnetic beads in a simple mix-and-go format for use in custom affinity purification experiments.
  • High Capacity—at least four times higher binding capacity than NHS-Activated Magnetic Beads from other suppliers
  • Easy to use—Immobilise in a simple one-step reaction with minimal hands-on time
  • Safe—No hazardous chemicals needed (e.g., Sodium Cyanoborohydride and Cyanogen Bromide)
  • Ligand-Compatible—use to immobilise with nearly any primary amine-containing compound or affinity ligand
  • Low non-specific binding—the bead surface is pre-blocked and any nonreacted NHS-Ester groups are fully quenched
  • Protocol-compatible—Protein coupling to the beads and downstream applications can be performed manually or by automation (e.g., Thermo Scientific KingFisher Instruments)
The activated magnetic beads contain N-hydroxy-succinimide (NHS) functional groups that react with primary amines forming stable amide linkages.
Pierce NHS-Activated Magnetic Beads offer a convenient way to conjugate any desired protein to a magnetic bead surface. The process does not require hazardous chemicals or lengthy reaction schemes. The beads are first incubated with protein for 1 to 2 hours in an amine free buffer at pH 7 to 9 to allow for covalent coupling through NHS-ester chemistry. The beads are subsequently washed and then any remaining active NHS-ester groups are quenched. The amine reactive chemistry and subsequent quench are completed in 3 to 4 hours. Coupled protein does not leach from the bead surface. In addition, the beads exhibit very low non-specific binding due to effective quenching and a proprietary blocking agent that coats the surface of the base particle. Following conjugation the prepared magnetic beads are typically used in affinity purification procedures.
Once they are covalently attached, the immobilised proteins are highly resistant to leaching from the bead surface. When prepared beads are used in experiments, nonspecific binding is negligible because nonreacted NHS-ester groups are thoroughly blocked during the coupling procedure. Pierce NHS-Activated Magnetic beads can be coupled and processed either manually with a magnetic stand or with automated platforms such as the Thermo Scientific KingFisher Instruments.
Bestellinformation: Upon receipt store at 4 °C. Product shipped with an ice pack.
Achtung! Bitte beachten: For Research Use Only. Not for use in diagnostic procedures.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge