Direct magnetic IP/Co-IP kit, Pierce™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
88828
PIER88828EA 614 EUR InStock
PIER88828
Direct magnetic IP/Co-IP kit, Pierce™
Protein-Aufbereitungs Protein Purification Kits
The Direct Magnetic IP/Co-IP Kit uses NHS-chemistry to covalently immobilise IP antibodies onto magnetic beads for effective immunoprecipitation and co-immunoprecipitation.
  • Complete antibody conjugation and immunoprecipitation in less than 4 hours
  • Use with any antibody species, class or isotype
  • Low non specific binding – bead surface is pre-blocked and all non reacted NHS-ester groups are fully quenched
  • Protein coupling to the beads and immunoprecipitation can be performed manually or by automation
The activated magnetic beads contain N-hydroxysuccinimide (NHS) functional groups, which react with primary amines on antibodies to form stable amide linkages. The antibody is first coupled to the NHS-activated magnetic beads in an amine-free buffer. The antibody-coupled beads are washed to remove any non-covalently bound antibody and quenched. The prepared beads are incubated with the antigen sample, washed to remove non-bound material
and eluted in a low-pH elution buffer to dissociate the antigen. The kit includes optimised buffers that minimise non specific binding and maximise antigen yield. Lane marker sample buffer is included for preparing samples for SDS-PAGE analysis. The beads are manually removed from the solution with a magnetic stand or by automation.
Lieferumfang: The kit includes NHS-activated magnetic beads, 1 ml; IP lysis/wash buffer, 2×50 ml; elution buffer (pH 2,0), 5 ml; lane marker sample buffer, non-reducing, (5X), 5 ml; neutralisation buffer (pH 8,5), 0,5 ml; 0,67 M borate buffer, 1 ml, BupH borate buffer pack, 1 pack; quenching buffer, 25 ml.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge