Anti-HA magnetic beads, Pierce™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
88836 88837
PIER88836EA 483 EUR InStock
PIER88836 PIER88837
Anti-HA magnetic beads, Pierce™
Kugeln Magnetkugeln
The Pierce™ Anti-HA magnetic beads are affinity particles for immunoprecipitation of recombinant HA-tagged proteins expressed in bacterial or mammalian cells or in vitro systems, using manual or robotic magnetic separators.
  • Highly specific anti-HA monoclonal antibody enables high yield and high purity immunoprecipitation
  • Convenient and fast with low non specific binding
  • Beads are compatible with manual and automated workflows
Stable, pre-blocked beads and specific antibody minimize off-target binding for HA-tag IP or co-IP experiments
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge