Protein L magnetic beads, Pierce™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
88850 88849
PIER88850EA 912 EUR InStock
PIER88850 PIER88849
Protein L magnetic beads, Pierce™
Kugeln Magnetkugeln
These protein L magnetic beads are high-capacity and high-throughput affinity particles for antibody purification using manual or robotic magnetic separators.
  • Immobilized Protein L is ideal for selective purification of human and mouse antibodies that have kappa light chains
  • Low non-specific binding of stable, pre-blocked beads provide clean purification of antibody
  • Beads are compatible with manual and automated workflows making it versatile
Recombinant Protein L, covalently attached to a blocked magnetic bead surface, selectively binds mouse and human antibodies through kappa light chains. The beads are commonly used to purify monoclonal antibodies in cell culture supernatants supplemented with bovine serum as Protein L does not bind bovine IgG.
These beads are ideal for purification of monoclonal and polyclonal antibodies that bind poorly to Protein A or Protein G magnetic beads, purification of monoclonal antibodies from culture supernatants supplemented with bovine serum, protein L IP and co-IP assays and purification of ScFv and Fab fragments containing kappa light chains.
They are typically used for purifying mouse and human antibodies containing kappa light chains from serum, cell culture supernatant or ascites. Protein L can bind a broader range of Ig classes including IgG, IgM, IgA, IgE and IgD. It also binds strongly to human (kappa I, III and IV only), mouse (kappa I only), rat and pig immunoglobulins.
It binds weakly to rabbit immunoglobulins and does not bind bovine, goat or sheep immunoglobulins. Single chain variable fragments (scFv) and Fab fragments also bind to Protein L.
The beads can be used manually with a magnetic stand, as well as with automated platforms.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge