Pierce™, NAb™ Affinity Purification Kits, Spin Kits for Antibody Purification
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
89951 89978 89981 89948 89949 89950 89952 89953 89954 89955 89956 89957 89958 89959 89960 89961 89962 89963 89979 89980
PIER89951EA 494 EUR InStock
PIER89951 PIER89978 PIER89981 PIER89948 PIER89949 PIER89950 PIER89952 PIER89953 PIER89954 PIER89955 PIER89956 PIER89957 PIER89958 PIER89959 PIER89960 PIER89961 PIER89962 PIER89963 PIER89979 PIER89980
Pierce™, NAb™ Affinity Purification Kits, Spin Kits for Antibody Purification
Protein-Aufbereitungs Protein Purification Kits
The Pierce™ NAb™ Spin Kits are convenient for rapid, small-scale affinity purification of antibodies from a variety of sample types. Each pre-filled microcentrifuge spin column of the immobilised protein resin enables quick purification of 100 to 1000 µg of IgG from 25 to 500 µl of serum or other sample. The actual amount of IgG purified varies depending upon the sample type and the specific spin column used.
  • Resin support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Superior manufacturing method immobilises by charge-free, leach-resistant covalent bonds, resulting in low non specific binding and enabling multiple uses without decline in yield
Proteins A and G bind to many of the same species and subclasses of IgG, although they have particular differences in affinity and binding capacity. Protein A is generally preferred for affinity purification of rabbit, pig, dog and cat IgG. Protein G has better binding capacity for a broader range of mouse and human IgG subclasses (IgG1, IgG2, etc.). Protein A/G is a recombinant fusion protein that includes the IgG-binding domains of both Protein A and Protein G. Therefore, Protein A/G is ideal for binding the broadest range of IgG subclasses from rabbit, mouse, human and other mammalian samples.
Protein L binds to certain immunoglobulin kappa light chains. Because kappa light chains occur in members of all immunoglobulin classes (i.e., IgG, IgM, IgA, IgE and IgD), Protein L can purify these different classes of antibody. However, only those antibodies within each class that possess the appropriate kappa light chains will bind. Generally, empirical testing is required to determine if Protein L is effective for purifying a particular antibody.
Lieferumfang: Each NAb™ Spin Kit, 0,2 ml contains spin columns (10), microcentrifuge tubes, 2 ml (80), Binding Buffer (50 ml), IgG Elution Buffer (1 pack, 500 ml) and Neutralisation Buffer (12 ml). Spin columns also available separately. Each NAb™ Spin Kit, 1 ml, contains spin columns (2), Binding Buffer (1 pack, 500 ml), IgG Elution Buffer (240 ml) and Neutralisation Buffer (12 ml).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge