Rapid equilbrium dialysis (RED) device, Pierce™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
89811 90006 90007 90085 90087 90088 99006 90112 91012
PIER89811EA 550 EUR InStock
PIER89811 PIER90006 PIER90007 PIER90085 PIER90087 PIER90088 PIER99006 PIER90112 PIER91012
Rapid equilbrium dialysis (RED) device, Pierce™
Röhrchen Dialyseröhrchen
Single-Use RED (rapid equilibrium dialysis) Plate is composed of disposable high-density polypropylene and is pre-loaded with 48 equilibrium dialysis membrane inserts. Each insert is comprised of two side-by-side chambers separated by an O-ring-sealed vertical cylinder of dialysis membrane with varying molecular-weight cutoffs (8 or 12 kD MWCO).
  • Easy and ready to use
  • Design enables equilibrium to reach 200 min⁻¹ agitation with in three to four hours
  • Reproducible and accurate for plasma binding assays
  • Robust compartmentalised design eliminates potential for leakage
  • For guaranteed performance each lot of inserts is functionally tested in a protein-binding assay
RED device is specifically designed and extensively validated for plasma serum binding assays. In addition to plasma protein binding, the device is used for determining drug partition between red blood cell and plasma; protein binding of liver microsomes to improve the correlation between in vitro and in vivo intrinsic clearance; and the competition between tissue protein binding against plasma proteins. The single-use RED plate is validated for minimal nonspecific binding.
This format requires no extensive assembly steps or specialised equipment, and each chamber/well is easily accessible from the top of the device. The base plate has a standard 96-well plate footprint with 9×9 mm well spacing. Additionally, the high membrane surface-to-volume ratio allows rapid dialysis, where equilibrium can be reached in 4 hours with high levels of reproducibility and accuracy.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge